Sodium hydroxide 4% (w/v) in aqueous solution (40 g/L) analytical reagent

Supplier: Ricca Chemical
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

R7247000-4A R7247000-500A R7247000-20F R7247000-1A
RC72471EA 153 USD
RC72471 RC724716 RC7247-5 RC7247-32
Sodium hydroxide 4% (w/v) in aqueous solution (40 g/L) analytical reagent
Sodium hydroxide
Formula: NaOH
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 4.00 ± 0.04 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR