Sodium chloride 32.5% (w/v) saturated aqueous solution

Supplier: Ricca Chemical

R7220500-1A
RC7220.5-32EA 131 USD
RC7220.5-32
Sodium chloride 32.5% (w/v) saturated aqueous solution
Sodium chloride
Formula: NaCl
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Specific Gravity Minimum 1.195 at 20 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR