Sodium chloride 24% (w/v) in hydrochloric acid 2%

Supplier: Ricca Chemical
Danger

R7219000-500A
RC721916EA 99 USD
RC721916
Sodium chloride 24% (w/v) in hydrochloric acid 2%
Sodium chloride
Formula: NaCl
Boiling Pt: ~ 105 °C
Density: 1.1580 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Refractive Index Confirmed Concentration


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR