Sodium chloride 0.9% (w/v) in aqueous solution

Supplier: Ricca Chemical

R7211000-20F
RC7211-5EA 126 USD
RC7211-5
Sodium chloride 0.9% (w/v) in aqueous solution
Sodium chloride

pH 7.25 --> 7.35 (25°C)

Formula: NaCl
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 0.899 ± 0.001 % (w/v)
pH at 25 °C (traceable to NIST SRM 186 & 191) 7.30 ± 0.02 pH at 25.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR