Sodium carbonate 10% (w/v) in aqueous solution analytical reagent
Supplier: Ricca Chemical
Synonyms:
Soda, Carbonic acid sodium salt, Calcined soda, Disodium carbonate, Carbonic acid disodium salt, Na-X, Bisodium carbonate, Carbonic acid, disodium salt, Soda ash
R7160000-4A
R7160000-500A
R7160000-1A
RC71601EA
141
USD
RC71601
RC716016
RC716032
Sodium carbonate 10% (w/v) in aqueous solution analytical reagent
Sodium carbonate
Formula:
Na₂CO₃ MW: 105.99 g/mol Boiling Pt: 1600 Melting Pt: 854 Storage Temperature: Ambient |
MDL Number:
MFCD00003494 CAS Number: 497-19-8 EINECS: 207-838-8 Merck Index: 14,08596 |
|
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay | 10.0 ± 0.3 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...