Sodium bromide 10% (w/v) in aqueous solution

Supplier: Ricca Chemical

R7146000-4A
RC7146-1EA 439 USD
RC7146-1
Sodium bromide 10% (w/v) in aqueous solution
Sodium bromide
Formula: NaBr
MW: 102.89 g/mol
Boiling Pt: ~ 100 °C
Density: 1.0610 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003475
CAS Number: 7647-15-6

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Refractive Index 105 ± 5 g/L

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR