Sodium azide 5% (w/v) in aqueous solution

Supplier: Ricca Chemical

Synonyms: Sodium triazide

7144.8-16
RC7144.8-16EA 91 USD
RC7144.8-16
Sodium azide 5% (w/v) in aqueous solution
Sodium azide
Formula: NaN₃
MW: 65.01 g/mol
MDL Number: MFCD00003536
CAS Number: 26628-22-8
UN: 1687
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR