Simulated Intestinal Fluid (without Pancreatin), USP XXII Formulation, RICCA Chemical Company
Supplier: Ricca Chemical
R7109750-500A
RC7109.75-16EA
104
USD
RC7109.75-16
Simulated Intestinal Fluid (without Pancreatin), USP XXII Formulation, RICCA Chemical Company
Standards
Biological Standards
Appearance: Colorless liquid
pH at 25°C (Method: SQCP027, SQCP033) 7.40-7.60
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...