Silver nitrate 0.0100 N (N/100) in 2-propanol

Supplier: Ricca Chemical
Danger

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

R7026000-1B
RC7026-32EA 261 USD
RC7026-32
Silver nitrate 0.0100 N (N/100) in 2-propanol
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, alcohol odor
Assay at 20 °C (traceable to NIST SRM 999) 0.01000 ± 0.00005 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR