Silver nitrate 0.0141 M (0.0141 N) 1 mL = 0.5 mg Cl⁻

Supplier: Ricca Chemical

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

6860-16
RC6860-16EA 44 USD
RC6860-16
Silver nitrate 0.0141 M (0.0141 N) 1 mL = 0.5 mg Cl⁻
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
MDL Number: MFCD00003414
CAS Number: 7761-88-8
UN: 1493
ADR: 5.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 0.0141 ± 0.00002 N at 20 °C
NIST SRM 999

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR