Potassium thiocyanate 10% (w/v) in aqueous solution analytical reagent

Supplier: Ricca Chemical

Synonyms: Potassium rhodanide

R6555000-4A R6555000-500A R6555000-1A
RC6555-1EA 326 USD
RC6555-1 RC6555-16 RC6555-32
Potassium thiocyanate 10% (w/v) in aqueous solution analytical reagent
Potassium thiocyanate
Formula: KSCN
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Refractive Index Confirmed concentration


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR