Potassium peroxodisulfate 5% (w/v) in aqueous solution

Supplier: Ricca Chemical

R6450000-1A
RC6450-32EA 107 USD
RC6450-32
Potassium peroxodisulfate 5% (w/v) in aqueous solution
Potassium peroxodisulphate
Formula: K₂S₂O₈
MDL Number: MFCD00011386
CAS Number: 7727-21-1
UN: 1492
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR