Potassium permanganate 0.0200 M (0.100 N)

Supplier: Ricca Chemical

6400-16
RC6400-16EA 39 USD
RC6400-16
Potassium permanganate 0.0200 M (0.100 N)
Potassium permanganate
Formula: KMnO₄
MDL Number: MFCD00011364
CAS Number: 7722-64-7
UN: 1490
ADR: 5.1,II

Order Now

Specification Test Results

Assay 0.1 ± 0.0001 N at 20 °C
NIST SRM 40


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR