Potassium permanganate 5% (w/v) in aqueous solution low in mercury

Supplier: Ricca Chemical

Synonyms: Permanganic acid potassium salt

R6360000-500C R6360000-1C
RC636016EA 71 USD
RC636016 RC6360-32
Potassium permanganate 5% (w/v) in aqueous solution low in mercury
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear, dark purple, odorless
Assay 5.00 ± 0.08 % (w/v)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR