Potassium iodate 0.0250 N (N/40)

Supplier: Ricca Chemical

R6273000-1C
RC6273-32EA 93 USD
RC6273-32
Potassium iodate 0.0250 N (N/40)
Potassium iodate
Formula: KlO₃
MW: 214 g/mol
Melting Pt: 560
MDL Number: MFCD00011406
CAS Number: 7758-05-6
UN: 1479
ADR: 5.1 II
Merck Index: 13,07726

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.02500 ± 0.00001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR