Potassium iodate 0.00564 N

Supplier: Ricca Chemical

R6270000-1C
RC627032EA 133 USD
RC627032
Potassium iodate 0.00564 N
Potassium iodate

1 mL = 0.2 mg Cl2 200 ppm

Formula: KlO₃
MW: 214 g/mol
Melting Pt: 560
MDL Number: MFCD00011406
CAS Number: 7758-05-6
UN: 1479
ADR: 5.1 II
Merck Index: 13,07726

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.005640 ± 0.000001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR