Ammonium chloride 5% (w/v) in aqueous solution

Supplier: Ricca Chemical
Danger

Synonyms: Ammonium muriate, Salmiac

R0625000-500A
RC62516EA 76 USD
RC62516
Ammonium chloride 5% (w/v) in aqueous solution
Ammonium chloride
Formula: NH₄Cl
MW: 53.49 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0154 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Refractive Index 5.0 ± 0.2 % (w/v)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR