Potassium hydroxide 0.500 N (N/2) in aqueous solution

Supplier: Ricca Chemical
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

R6230000-4A R6230000-1A R6230000-20F
RC62301EA 215 USD
RC62301 RC623032 RC62305
Potassium hydroxide 0.500 N (N/2) in aqueous solution
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 0.5000 ± 0.0002 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR