Ammonium carbonate USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

R0622500-500A
RC622.5-16EA 166 USD
RC622.5-16
Ammonium carbonate USP Test Solution (TS)
Ammonium carbonate
Formula: (NH₄)₂CO₃
Density: 1.1000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010890
CAS Number: 506-87-6

Order Now

Specification Test Results

Appearance Clear, colorless, ammonia odor
Assay at 20 °C (traceable to NIST SRM 723) 4.73 to 5.19 N Ammonia


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR