Potassium fluoride 40% (w/w) in aqueous solution

Supplier: Ricca Chemical

R6104000-20F
RC6104-5EA 1382 USD
RC6104-5
Potassium fluoride 40% (w/w) in aqueous solution
Potassium fluoride
Formula: KF
MDL Number: MFCD00011398
CAS Number: 7789-23-3
UN: 1812
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear, light pink
Specific Gravity 1.40 ± 0.01 at 25.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR