Potassium dichromate 0.200 N (N/5) in aqueous solution

Supplier: Ricca Chemical
Danger

R6065000-1A
RC6065-32EA 115 USD
RC6065-32
Potassium dichromate 0.200 N (N/5) in aqueous solution
Potassium dichromate
Formula: K₂Cr₂O₇
Storage Temperature: Ambient
MDL Number: MFCD00011367
CAS Number: 7778-50-9
EINECS: 231-906-6
UN: 3287
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear, orange, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.2000 ± 0.0001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR