Potassium dichromate 106 g/L solution R

Supplier: Ricca Chemical
Danger

R6044000-500A
RC6044-16EA 131 USD
RC6044-16
Potassium dichromate 106 g/L solution R
Potassium dichromate
Formula: K₂Cr₂O₇
Storage Temperature: Ambient
MDL Number: MFCD00011367
CAS Number: 7778-50-9
EINECS: 231-906-6
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear, orange, odorless
Refractive Index 106 ± 2 g/L


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR