Potassium carbonate 7% (w/v) in aqueous solution USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

R5882500-500A
RC5882.5-16EA 133 USD
RC5882.5-16
Potassium carbonate 7% (w/v) in aqueous solution USP Test Solution (TS)
Potassium carbonate
Formula: K₂CO₃
Boiling Pt: ~ 100 °C
Density: 1.0604 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 7.00 ± 0.02 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR