Potassium bromide 10% (w/v) in aqueous solution

Supplier: Ricca Chemical
Warning

R5879500-4A R5879500-1A
RC5879.5-1EA 553 USD
RC5879.5-1 RC5879.5-32
Potassium bromide 10% (w/v) in aqueous solution
Potassium bromide
Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435
Melting Pt: 730
Density: 1.0708 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
Merck Index: 13,07701

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 10.0 ± 0.1 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR