Phosphoric acid - sulfamic acid solution for residual chlorine determination
Supplier: Ricca Chemical
|
Danger
|
R5854000-500A
RC585416EA
85
USD
RC585416
Phosphoric acid - sulfamic acid solution for residual chlorine determination
Phosphoric acid - sulfamic acid solution
Boiling Pt:
~ 105 °C Density: 1.0890 g/cm³ Storage Temperature: Ambient |
UN:
1805 ADR: 8,III |
Specification Test Results
Appearance | Clear, colorless, odorless |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...