Orthophosphoric acid 0.1% (v/v) in aqueous solution

Supplier: Ricca Chemical

Synonyms: Phosphoric acid

R5845000-4A
RC5845-1EA 91 USD
RC5845-1
Orthophosphoric acid 0.1% (v/v) in aqueous solution
Orthophosphoric acid
Formula: H₃PO₄
MW: 98 g/mol
MDL Number: MFCD00011340
CAS Number: 7664-38-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 0.100 ± 0.002 % (v/v)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR