Phosphate Standard, 1 ml = 0.1 mg PO₄³⁻, 100 ppm PO₄³⁻ (32.6 ppm P), Ricca Chemical Company
Supplier: Ricca Chemical
R5833000-120A
RC5833-4EA 50 USD
RC5833-4
Phosphate Standard, 1 ml = 0.1 mg PO₄³⁻, 100 ppm PO₄³⁻ (32.6 ppm P), Ricca Chemical Company
Standards Anionic Standards Phosphate Standards