Aluminium hydroxide, suspension for chloride determination in highly colored samples

Supplier: Ricca Chemical
Warning

R0580000-500A
RC58016EA 151 USD
RC58016
Aluminium hydroxide, suspension for chloride determination in highly colored samples
Aluminium hydroxide
Formula: Al(OH)₃
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.1000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003420
CAS Number: 21645-51-2

Order Now

Specification Test Results

Appearance Opaque, white suspension, ammonia odor
Chloride Free Check Chloride Free


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR