Phenolphthalein 1% (w/v) in 2-propanol 60% (v/v)/water 40% (v/v), neutralized

Supplier: Ricca Chemical
Danger

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

5625.6-16 5625.6-4
RC5625.6-16EA 113 USD
RC5625.6-16 RC5625.6-4
Phenolphthalein 1% (w/v) in 2-propanol 60% (v/v)/water 40% (v/v), neutralized
Phenolphthalein
Formula: C₂₀H₁₄O₄
Boiling Pt: ~ 82 °C
Melting Pt: ~ 0 °C
Density: 0.8680 g/cm³
Flash Pt: ~ 13 °C (IPA)
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Suitability Colorless (pH 8.0) - Pink (pH 10.0)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR