Phenolphthalein 1% (w/v) in 2-propanol 60% (v/v)/water 40% (v/v), neutralized
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
5625.6-16
5625.6-4
RC5625.6-16EA
113
USD
RC5625.6-16
RC5625.6-4
Phenolphthalein 1% (w/v) in 2-propanol 60% (v/v)/water 40% (v/v), neutralized
Phenolphthalein
Formula:
C₂₀H₁₄O₄ Boiling Pt: ~ 82 °C Melting Pt: ~ 0 °C Density: 0.8680 g/cm³ Flash Pt: ~ 13 °C (IPA) Storage Temperature: Ambient |
MDL Number:
MFCD00005913 CAS Number: 77-09-8 UN: 1987 ADR: 3,III |
|
Specification Test Results
Suitability | Colorless (pH 8.0) - Pink (pH 10.0) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...