Oxalic acid stock solution 10 g/L

Supplier: Ricca Chemical
Danger

R5471000-500A
RC5471-16EA 120 USD
RC5471-16
Oxalic acid stock solution 10 g/L
Oxalic acid
Formula: C₂H₂O₄
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0034 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS Number: 144-62-7
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 40) 10.0 ± 0.3 g/L


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR