Oxalic acid 6.3% (w/v) in aqueous solution USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

R5466000-1A R5466000-500A
RC5466-32EA 85 USD
RC5466-32 RC5466-16
Oxalic acid 6.3% (w/v) in aqueous solution USP Test Solution (TS)
Oxalic acid
Formula: C₂H₂O₄
MDL Number: MFCD00002573
CAS Number: 144-62-7
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Refractive Index 6.3 ± 0.5 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR