Nitrogen Standard, 1 ml = 0.25 mg N, 250 ppm N as Nitrite (821 ppm NO₂⁻), Equivalent to 1 mL = 821.1 ppm Nitrite (NO₂), Ricca Chemical Company
Supplier: Ricca Chemical
R5460000-500A R5460000-120A
RC546016EA 187 USD
RC546016 RC54604
Nitrogen Standard, 1 ml = 0.25 mg N, 250 ppm N as Nitrite (821 ppm NO₂⁻), Equivalent to 1 mL = 821.1 ppm Nitrite (NO₂), Ricca Chemical Company
Standards Anionic Standards Nitrite Standards
APHA for nitrite
250 ppm nitrite.