Nitrogen Standard, 1 ml = 0.1 mg N, 100 ppm N as Nitrate (442.7 ppm NO₃⁻), Ricca Chemical Company
Supplier: Ricca Chemical
R5457000-500A
RC545716EA 94 USD
RC545716
Nitrogen Standard, 1 ml = 0.1 mg N, 100 ppm N as Nitrate (442.7 ppm NO₃⁻), Ricca Chemical Company
Standards Anionic Standards Nitrate Standards
APHA for nitrate
100 ppm nitrogen.