Nitrite Standard, 1 ml = 0.200 mg NO₂⁻, 200 ppm NO₂⁻ (60.9 ppm N), Ricca Chemical Company
Supplier: Ricca Chemical
R5444200-120C
RC5444.2-4EA 90 USD
RC5444.2-4
Nitrite Standard, 1 ml = 0.200 mg NO₂⁻, 200 ppm NO₂⁻ (60.9 ppm N), Ricca Chemical Company
Standards Anionic Standards Nitrite Standards