Nitrate Standard, 1 m = 1 mg NO₃⁻, 1000 ppm NO₃⁻ (226 ppm N), Ricca Chemical Company
Supplier: Ricca Chemical
R5307000-120A
R5307000-500A
RC5307-4EA
54
USD
RC5307-4
RC5307-16
Nitrate Standard, 1 m = 1 mg NO₃⁻, 1000 ppm NO₃⁻ (226 ppm N), Ricca Chemical Company
Standards
Anionic Standards
Nitrate Standards