N-(1-Naphthyl)ethylenediamine dihydrochloride USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

Synonyms: Marshall's Reagent, N-2-Aminoethyl-1-naphthylamine dihydrochloride, N-1-Naphthylethylenediamine

R5231000-500C
RC5231-16EA 88 USD
RC5231-16
N-(1-Naphthyl)ethylenediamine dihydrochloride USP Test Solution (TS)
N-2-Aminoethyl-1-naphthylamine dihydrochloride
Formula: C₁₂H₁₆Cl₂N₂
MW: 259.18 g/mol
Boiling Pt: ~ 60 °C
Density: 0.8526 g/cm³
Flash Pt: –18 °C (Acetone)
Storage Temperature: Refrigerator
MDL Number: MFCD00012556
CAS Number: 1465-25-4
UN: 1090
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3643e6df3e9cf8dc627823e9090c97004ee25308&vid=a30fc28279c11af12c6b735b00b84fc8f08caf34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, greenish-yellow to slight brown, Acetone odor

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR