N-(1-Naphthyl)ethylenediamine dihydrochloride 0.1% (w/v) in aqueous solution

Supplier: Ricca Chemical

Synonyms: Marshall's Reagent, N-2-Aminoethyl-1-naphthylamine dihydrochloride, N-1-Naphthylethylenediamine

R5230000-500C
RC523016EA 115 USD
RC523016
N-(1-Naphthyl)ethylenediamine dihydrochloride 0.1% (w/v) in aqueous solution
N-2-Aminoethyl-1-naphthylamine dihydrochloride
Formula: C₁₂H₁₆Cl₂N₂
MW: 259.18 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0050 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00012556
CAS Number: 1465-25-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3643e6df3e9cf8dc627823e9090c97004ee25308&vid=a30fc28279c11af12c6b735b00b84fc8f08caf34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, greenish-yellow to slight brown, odorless

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR