Methylene blue (Loeffler) for staining and differentiation of acid fast organisms in smears

Supplier: Ricca Chemical
Danger

Synonyms: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue

R4880000-500A R4880000-120A
RC488016EA 72.87 USD
RC488016 RC48804
Methylene blue (Loeffler) for staining and differentiation of acid fast organisms in smears
Methylene blue

Methylene blue, Loeffler formulation

Formula: C₁₆H₁₈ClN₃S
MW: 319.86 g/mol
Boiling Pt: ~ 82 °C
Melting Pt: 180
Density: 0.9800 g/cm³
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00150008
CAS Number: 61-73-4
UN: 1987
ADR: 3 III
Merck Index: 12,06137

Order Now

Specification Test Results

Absorbance (traceable to NIST SRM 930) Conforms to control
Appearance Dark blue, slight alcohol odor


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR