Methylene blue 1% (w/v) in denatured alcohol
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue
R4864000-120A
R4864000-500A
RC4864-4EA
51
USD
RC4864-4
RC4864-16
Methylene blue 1% (w/v) in denatured alcohol
Methylene blue
Formula:
C₁₆H₁₈ClN₃S MW: 319.86 g/mol Boiling Pt: ~ 77 °C Melting Pt: 180 Density: 0.8000 g/cm³ Flash Pt: ~ 15.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150008 CAS Number: 61-73-4 UN: 1987 ADR: 3 II Merck Index: 12,06137 |
Specification Test Results
Absorbance (traceable to NIST SRM 930) | Conforms to control |
Appearance | Dark blue, alcohol odor |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...