Mercury(II) acetate 6% (w/v) in glacial acetic acid USP Test Solution (TS) analytical reagent

Supplier: Ricca Chemical
Danger

Synonyms: Mercury diacetate, Mercuric acetate, Acetic acid mercury (II) salt

R4629000-1C R4629000-500C
RC4629-32EA 488 USD
RC4629-32 RC4629-16
Mercury(II) acetate 6% (w/v) in glacial acetic acid USP Test Solution (TS) analytical reagent
Mercury (II) acetate

Clear, colorless liquid with a strong vinegar odor.

Formula: (H₃CCOO)₂Hg
Melting Pt: 178…180 °C
Density: 3.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012165
CAS Number: 1600-27-7
EINECS: 216-491-1
UN: 1629
ADR: 6.1,II
Merck Index: 13,05898

Order Now

Specification Test Results

Appearance Clear, colorless, strong acetic odor


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR