Magnesium sulfate 12% (w/v) in aqueous solution USP Test Solution (TS)
Supplier: Ricca Chemical
Synonyms:
MgSO4, Magnesium sulfate anhydrous, Magnesium sulphate anhydrous
R4548000-500A
R4548000-1A
RC4548-16EA
49
USD
RC4548-16
RC4548-32
Magnesium sulfate 12% (w/v) in aqueous solution USP Test Solution (TS)
Magnesium sulphate
Formula:
MgSO₄ MW: 120.37 g/mol Boiling Pt: ~ 100 °C Melting Pt: 1127 Density: 1.0570 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011110 CAS Number: 7487-88-9 Merck Index: 12,05731 |
|
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 915) | 12.0 ± 0.1 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...