Lactic acid 0.100 N (N/10)

Supplier: Ricca Chemical

R4275000-500A
RC427516EA 63 USD
RC427516
Lactic acid 0.100 N (N/10)
Lactic acid
Formula: H₃CCH(OH)COOH
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0018 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 0.1000 ± 0.0001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR