2-Propanol 70% (v/v)

Supplier: Ricca Chemical
Danger

R4210000-55E R4210000-20E1 R4210000-20F R4210000-4A R4210000-4C1
RC4210-55EA 2585 USD
RC4210-55 RC4210-5HP RC42105 RC42101 RCR4210000-4C1
2-Propanol 70% (v/v)
2-Propanol
Formula: (CH₃)₂CHOH
Boiling Pt: ∼ 82°C
Melting Pt: ∼ 0°C
Density: 0.8770
Flash Pt: ∼ 13°C (55°F) (100%); 18°C (64°F) (70%)
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1219
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (by Specific Gravity) 68-72 % (v/v)
Specific Gravity at 20°C 0.874-0.882


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR