Hydrochloric acid 0.100 N (N/10) in 2-propanol

Supplier: Ricca Chemical
Danger

R3800000-4A R3800000-1A R3800000-20F R3800000-500A
RC38001EA 338 USD
RC38001 RC380032 RCR3800000-20F RCR3800000500A
Hydrochloric acid 0.100 N (N/10) in 2-propanol
Hydrochloric acid
Formula: HCl
Density: 0.79 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1219
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 0.0995-0.1005 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR