Hydrochloric acid 6.00 N

Supplier: Ricca Chemical
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

R3750100-1C
RC3750.1-32EA 82 USD
RC3750.1-32
Hydrochloric acid 6.00 N
Hydrochloric acid

Packaged in Glass

Formula: HCl
MW: 36.46 g/mol
Density: 1.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless to slight yellow-green, Hydrochloric odor
Assay at 20 °C (traceable to NIST SRM 84) 6.000 ± 0.005 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR