Hydrochloric acid 5.00 N

Supplier: Ricca Chemical

Synonyms: Hydrochloric acid solution, Muriatic acid

3740-32G
RC3740-32GEA 58 USD
RC3740-32G
Hydrochloric acid 5.00 N
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Assay 5 ± 0.005 N at 20 °C
NIST SRM 84

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR