Hydrochloric acid 0.100 N (N/10)

Supplier: Ricca Chemical
Danger

R3600000-4A R3600000-1A R3600000-20F R3600000-500A R3600000-20E1 R3600000-10F
RC36001EA 110 USD
RC36001 RC360032 RC36005 RC3600-16 RC3600-5HP RCR3600000-10F
Hydrochloric acid 0.100 N (N/10)
Hydrochloric acid

Standardized at 25 °C against chemicals whose certification is traceable to NIST.

Formula: HCl
Density: 1.004 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Tris(hydroxymethyl)aminomethane/Methyl Red) 0.0999-0.1001 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR