Sulfuric acid solution alcoholic solution for phosphorus determination

Supplier: Ricca Chemical

R0350000-1A
RC35032EA 132 USD
RC35032
Sulfuric acid solution alcoholic solution for phosphorus determination
Sulfuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, alcohol odor

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR