Glycine 10% (w/v) in aqueous solution

Supplier: Ricca Chemical

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

R3283000-500A
RC3283-16EA 77 USD
RC3283-16
Glycine 10% (w/v) in aqueous solution
Glycine
Formula: NH₂CH₂COOH
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0500 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, odorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR